6-Bromo-1,4-dihydro-2H-3,1-benzoxazin-2-one 95%
Supplier: Apollo Scientific
Warning
OR400397-1G OR400397-5G
APOSOR400397-1GEA 66.7 EUR
APOSOR400397-1G APOSOR400397-5G
6-Bromo-1,4-dihydro-2H-3,1-benzoxazin-2-one 95%
6-Bromo-1,4-dihydro-2H-3,1-benzoxazin-2-one
Formula: C₈H₆BrNO₂
MW: 228.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972273
CAS Number: 1017783-09-3