2-(Hydroxymethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
Warning
OR40041-25G OR40041-5G
APOSOR40041-25GEA 31.6 EUR
APOSOR40041-25G APOSOR40041-5G
2-(Hydroxymethyl)benzyl alcohol 98%
2-(Hydroxymethyl)benzyl alcohol
Formula: C₈H₁₀O₂
Melting Pt: 59…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9