Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate 95+%
Supplier: Apollo Scientific
|
Warning
|
OR40042-1G
OR40042-5G
OR40042-25G
APOSOR40042-1GEA
21.9
EUR
APOSOR40042-1G
APOSOR40042-5G
APOSOR40042-25G
Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate 95+%
Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate
Formula:
C₉H₉NO₆ Melting Pt: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203667 CAS Number: 42590-00-1 |