2-Bromo-5-iodobenzoic acid
Supplier: Apollo Scientific
Warning
OR4005-1G OR4005-25G OR4005-5G
APOSOR4005-1GEA 21.9 EUR
APOSOR4005-1G APOSOR4005-25G APOSOR4005-5G
2-Bromo-5-iodobenzoic acid
2-Bromo-5-iodobenzoic acid
Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00079716
CAS Number: 25252-00-0