(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide 95%
Supplier: Apollo Scientific
Warning
Synonyms: (S)-Tetrahydro-3H-pyrrolo[1,2-c][1,2,3]oxathiazole 1,1-dioxide
OR400820-1G OR400820-250MG OR400820-5G
APOSOR400820-1GEA 36.4 EUR
APOSOR400820-1G APOSOR400820-250MG APOSOR400820-5G
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide 95%
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide
Formula: C₅H₉NO₃S
MW: 163.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123262
CAS Number: 132635-95-1