4-Bromo-3-iodoaniline 95+%
Supplier: Apollo Scientific
|
Danger
|
OR400870-250MG
OR400870-1G
OR400870-5G
APOSOR400870-250MGEA
89.9
EUR
APOSOR400870-250MG
APOSOR400870-1G
APOSOR400870-5G
4-Bromo-3-iodoaniline 95+%
4-Bromo-3-iodoaniline
Formula:
C₆H₅BrIN MW: 297.9191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16622212 CAS Number: 63037-64-9 UN: 2811 ADR: 6.1,III |