2-Chloro-p-anisidine 97+%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-4-methoxyaniline
OR400894-100G OR400894-25G
APOSOR400894-100GEA 571 EUR
APOSOR400894-100G APOSOR400894-25G
2-Chloro-p-anisidine 97+%
2-Chloro-p-anisidine
Formula: C₇H₈ClNO
MW: 157.6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209639
CAS Number: 29242-84-0