2-Chloro-5-methoxybenzaldehyde
Supplier: Apollo Scientific
OR400965-5G
APOSOR400965-5GEA 114 EUR
APOSOR400965-5G
2-Chloro-5-methoxybenzaldehyde
2-Chloro-5-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845925
CAS Number: 13719-61-4