3-Chloro-5-methoxybenzonitrile, Technical Grade
Supplier: Apollo Scientific
Warning
OR401002-1G OR401002-5G
APOSOR401002-1GEA 94.4 EUR
APOSOR401002-1G APOSOR401002-5G
3-Chloro-5-methoxybenzonitrile, Technical Grade
3-Chloro-5-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505060
CAS Number: 473923-96-5