3,6-Dichloropyridine-2-methanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3,6-Dichloro-2-(hydroxymethyl)pyridine
OR40359-1G OR40359-5G
APOSOR40359-1GEA 94.4 EUR
APOSOR40359-1G APOSOR40359-5G
3,6-Dichloropyridine-2-methanol 95%
3,6-Dichloropyridine-2-methanol
Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Boiling Pt: 100…102 °C/0.2mmHg
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD13185536
CAS Number: 58804-10-7