(2-Amino-1,3-thiazol-5-yl)methanol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-(hydroxymethyl)-1,3-thiazole, (2-Aminothiazol-5-yl)methanol
OR40405-1G OR40405-250MG OR40405-5G
APOSOR40405-1GEA 227 EUR
APOSOR40405-1G APOSOR40405-250MG APOSOR40405-5G
(2-Amino-1,3-thiazol-5-yl)methanol
(2-Amino-1,3-thiazol-5-yl)methanol
Formula: C₄H₆N₂OS
MW: 130.17 g/mol
Boiling Pt: 340 °C/760mmHg
Melting Pt: 102…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203018
CAS Number: 131184-73-1