5-Chloro-2-nitrobenzyl alcohol
Supplier: Apollo Scientific
Warning
OR40411-10G OR40411-1G
APOSOR40411-10GEA 37.7 EUR
APOSOR40411-10G APOSOR40411-1G
5-Chloro-2-nitrobenzyl alcohol
5-Chloro-2-nitrobenzyl alcohol
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00007291
CAS Number: 73033-58-6
EINECS: 277-241-5