2-Bromo-m-cresol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-3-methylphenol
OR40426-25G OR40426-5G OR40426-1G
APOSOR40426-25GEA 302 EUR
APOSOR40426-25G APOSOR40426-5G APOSOR40426-1G
2-Bromo-m-cresol 98%
2-Bromo-m-cresol
Formula: C₇H₇BrO
MW: 187.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991797
CAS Number: 22061-78-5