2-Methoxy-4-nitrophenol 98%
Supplier: Apollo Scientific
Warning
OR40450-100G
APOSOR40450-100GEA 0 EUR
APOSOR40450-100G
2-Methoxy-4-nitrophenol 98%
2-Methoxy-4-nitrophenol
Formula: C₇H₇NO₄
Melting Pt: 99…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00012143
CAS Number: 3251-56-7