tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide
OR40453-25G OR40453-5G
APOSOR40453-25GEA 116 EUR
APOSOR40453-25G APOSOR40453-5G
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Formula: C₁₄H₂₂N₄O₄
MW: 310.354 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD02683427
CAS Number: 152120-54-2