1-Bromo-4-ethynylbenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne
OR40459-25G OR40459-5G
APOSOR40459-25GEA 89.9 EUR
APOSOR40459-25G APOSOR40459-5G
1-Bromo-4-ethynylbenzene 98%
1-Bromo-4-ethynylbenzene
Formula: C₈H₅Br
MW: 181.03 g/mol
Boiling Pt: 88…90 °C/16mmHg
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00168822
CAS Number: 766-96-1
UN: 3077
ADR: 9+EHS III