Bromoethylene 98%
Supplier: Apollo Scientific
|
Danger
|
APOSOR40494-2.2KGEA
1480
EUR
APOSOR40494-2.2KG
APOSOR40494-1KG
APOSOR40494-50G
APOSOR40494-250G
Bromoethylene 98%
Bromoethylene
Formula:
C₂H₃Br MW: 106,95 g/mol Boiling Pt: 16 °C Melting Pt: –139,5 °C Density: 1,517 g/cm³ (25 °C) Flash Pt: <–8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000183 EINECS: 209-800-6 UN: 1085 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...