1-Naphthylmethylamine 98%

Supplier: Apollo Scientific
Warning

OR40514-25G OR40514-5G
APOSOR40514-25GEA 166 EUR
APOSOR40514-25G APOSOR40514-5G
1-Naphthylmethylamine 98%
1-Naphthylmethylamine
Formula: C₁₁H₁₁N
Boiling Pt: 290…293 °C
Density: 1.073 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004048
CAS Number: 118-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR