Ethyl-4-methylthiazole-5-carboxylate 98%
Supplier: Apollo Scientific
Warning
OR40519-25G OR40519-5G
APOSOR40519-25GEA 56.7 EUR
APOSOR40519-25G APOSOR40519-5G
Ethyl-4-methylthiazole-5-carboxylate 98%
Ethyl-4-methylthiazole-5-carboxylate
Formula: C₇H₉NO₂S
Boiling Pt: 215…233 °C
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00626873
CAS Number: 20582-55-2