3-Methyl-2-nitrobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Methyl-2-nitrophenyl)methanol, 3-(Hydroxymethyl)-2-nitrotoluene
OR40576-1G OR40576-5G OR40576-25G
APOSOR40576-1GEA 29 EUR
APOSOR40576-1G APOSOR40576-5G APOSOR40576-25G
3-Methyl-2-nitrobenzyl alcohol 98%
3-Methyl-2-nitrobenzyl alcohol
Formula: C₈H₉NO₃
Melting Pt: 48…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007182
CAS Number: 80866-76-8