1,1,3-Trichloropropene 97%
Supplier: Apollo Scientific
Warning
OR40579-1G OR40579-5G
APOSOR40579-1GEA 323 EUR
APOSOR40579-1G APOSOR40579-5G
1,1,3-Trichloropropene 97%
1,1,3-Trichloropropene
Formula: C₃H₃Cl₃
MW: 145.41 g/mol
Boiling Pt: 135 °C
Density: 1.3953 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00013259
CAS Number: 2567-14-8
UN: 1993
ADR: 3,III