(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
Supplier: Apollo Scientific
Danger
Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-amine
OR40592-1G OR40592-25G OR40592-5G
APOSOR40592-1GEA 29 EUR
APOSOR40592-1G APOSOR40592-25G APOSOR40592-5G
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 250 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00671630
CAS Number: 23357-52-0
UN: 2735
ADR: 8,III