(S)-(-)-3-Butyne-2-ol 98%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-3-Butyn-2-ol, (S)-(-)-3-Hydroxy-1-butyne, (S)-(-)-But-3-yn-2-ol
OR40593-25G OR40593-5G
APOSOR40593-25GEA 306 EUR
APOSOR40593-25G APOSOR40593-5G
(S)-(-)-3-Butyne-2-ol 98%
(S)-(-)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 108…111 °C
Density: 0.88 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00190166
CAS Number: 2914-69-4
UN: 2929
ADR: 6.1+3,II