Coronene
Supplier: Apollo Scientific
Warning
Synonyms: Hexabenzobenzene
OR40595-1G OR40595-250MG
APOSOR40595-1GEA 220 EUR
APOSOR40595-1G APOSOR40595-250MG
Coronene
Coronene
Formula: C₂₄H₁₂
MW: 300.36 g/mol
Boiling Pt: 525 °C
Melting Pt: 428 °C
Storage Temperature: Ambient
MDL Number: MFCD00004134
CAS Number: 191-07-1
EINECS: 205-881-7