4,4'-Diphenoxybenzophenone 99%
Supplier: Apollo Scientific
Warning
Synonyms: Bis(4-phenoxyphenyl)methanone
OR40624-100G OR40624-25G OR40624-5G
APOSOR40624-100GEA 951 EUR
APOSOR40624-100G APOSOR40624-25G APOSOR40624-5G
4,4'-Diphenoxybenzophenone 99%
4,4'-Diphenoxybenzophenone
Formula: C₂₅H₁₈O₃
MW: 366.42 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00077969
CAS Number: 14984-21-5