4-iodobenzene-1,2-diamine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Iodo-o-phenylenediamine, 4-Iodobenzene-1,2-diamine
OR40649-5G
APOSOR40649-5GEA 111 EUR
APOSOR40649-5G
4-iodobenzene-1,2-diamine 97%
4-iodobenzene-1,2-diamine
Formula: C₆H₇IN₂
MW: 234.04 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669466
CAS Number: 21304-38-1