2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl 97%
Supplier: Apollo Scientific
Warning
Synonyms: XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl
OR40651-5G OR40651-25G OR40651-100G
APOSOR40651-5GEA 21.9 EUR
APOSOR40651-5G APOSOR40651-25G APOSOR40651-100G
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl 97%
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl
Formula: C₃₃H₄₉P
MW: 476.72 g/mol
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD04117682
CAS Number: 564483-18-7