2,3-Xylenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3-Dimethylphenol, 3-Hydroxy-2-xylene, 3-Hydroxy-o-xylene
OR40665-25G OR40665-5G OR40665-100G
APOSOR40665-25GEA 45.1 EUR
APOSOR40665-25G APOSOR40665-5G APOSOR40665-100G
2,3-Xylenol 98%
2,3-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 216…218 °C
Melting Pt: 70…76 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002227
CAS Number: 526-75-0
EINECS: 208-395-3
UN: 2928
ADR: 6.1+8 II
Merck Index: 12,10215