Triethylsilylacetylene 97%
Supplier: Apollo Scientific
Danger
OR40699-100G
APOSOR40699-100GEA 172 EUR
APOSOR40699-100G
Triethylsilylacetylene 97%
Triethylsilylacetylene
Formula: C₈H₁₆Si
MW: 140.3 g/mol
Density: 0.783 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00075373
CAS Number: 1777-03-3
UN: 1993
ADR: 3,III