Naphthalen-2-ylhydrazine hydrochloride 95+%
Supplier: Apollo Scientific
Warning
OR40713-100G
APOSOR40713-100GEA 58 EUR
APOSOR40713-100G
Naphthalen-2-ylhydrazine hydrochloride 95+%
Naphthalen-2-ylhydrazine hydrochloride
Formula: C₁₀H₁₁ClN₂
MW: 194.6607 g/mol
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD04109481
CAS Number: 2243-58-5