1,3-Dichloro-5,5-dimethylhydantoin
Supplier: Apollo Scientific
Synonyms: 1,3-Dichloro-5,5-dimethylhydantoine, 1,3-Dichloro-5,5-dimethylimidazolidine-2,4-dione
OR40788-100G OR40788-500G
APOSOR40788-100GEA 43.9 EUR
APOSOR40788-100G APOSOR40788-500G
1,3-Dichloro-5,5-dimethylhydantoin
1,3-Dichloro-5,5-dimethylhydantoin
Formula: C₅H₆Cl₂N₂O₂
MW: 197.02 g/mol
Melting Pt: 132…136 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00003190
CAS Number: 118-52-5
UN: 3085
ADR: 5.1+8 II