2-Bromo-4,5-dichloroimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4,5-dichloro-1H-imidazole
OR4087-25G OR4087-5G
APOSOR4087-25GEA 261 EUR
APOSOR4087-25G APOSOR4087-5G
2-Bromo-4,5-dichloroimidazole
2-Bromo-4,5-dichloroimidazole
Formula: C₃HBrCl₂N₂
MW: 215.86 g/mol
Melting Pt: 215…223 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00067894
CAS Number: 16076-27-0