1-Boc-4-iodopiperidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate
OR41125-25G OR41125-5G OR41125-100G
APOSOR41125-25GEA 55.1 EUR
APOSOR41125-25G APOSOR41125-5G APOSOR41125-100G
1-Boc-4-iodopiperidine 97%
1-Boc-4-iodopiperidine
Formula: C₁₀H₁₈INO₂
MW: 311.16 g/mol
Melting Pt: 47…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115041
CAS Number: 301673-14-3