1,3,5-Tribromobenzene
Supplier: Apollo Scientific
Warning
OR4123-100G OR4123-25G OR4123-500G
APOSOR4123-100GEA 42.2 EUR
APOSOR4123-100G APOSOR4123-25G APOSOR4123-500G
1,3,5-Tribromobenzene
1,3,5-Tribromobenzene
Formula: C₆H₃Br₃
Boiling Pt: 271 °C
Melting Pt: 117…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3