1-(1-Methylpiperidin-4-yl)piperazine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(1-Methyl-4-piperidinyl)piperazine
OR4165-1G OR4165-5G OR4165-250MG
APOSOR4165-1GEA 56.7 EUR
APOSOR4165-1G APOSOR4165-5G APOSOR4165-250MG
1-(1-Methylpiperidin-4-yl)piperazine 98%
1-(1-Methylpiperidin-4-yl)piperazine
Formula: C₁₀H₂₁N₃
MW: 183.3 g/mol
Boiling Pt: 112 °C/2mmHg
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD01075186
CAS Number: 23995-88-2
UN: 3259
ADR: 8 II