Ethyl-2-methylnicotinate 98%
Supplier: Apollo Scientific
Warning
OR42000-1G OR42000-5G
APOSOR42000-1GEA 21.9 EUR
APOSOR42000-1G APOSOR42000-5G
Ethyl-2-methylnicotinate 98%
Ethyl 2-methylnicotinate
Formula: C₉H₁₁NO₂
Boiling Pt: 126…127 °C/24mmHg
Density: 1.072 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006336
CAS Number: 1721-26-2