Methyl-1-boc-azetidine-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 1-Boc-azetidine-3-carboxylate, 1-tert-Butyl 3-methyl azetidine-1,3-dicarboxylate
OR42009-100G OR42009-25G OR42009-5G
APOSOR42009-100GEA 848 EUR
APOSOR42009-100G APOSOR42009-25G APOSOR42009-5G
Methyl-1-boc-azetidine-3-carboxylate
Methyl-1-boc-azetidine-3-carboxylate
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Boiling Pt: 265…272 °C
Density: 1.152 g/cm³ (25 °C)
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD06656775
CAS Number: 610791-05-4