4-Quinolinol 97%
Supplier: Apollo Scientific
Warning
Synonyms: Quinolin-4-ol, 4-Hydroxyquinoline
OR4202-100G
APOSOR4202-100GEA 79.9 EUR
APOSOR4202-100G
4-Quinolinol 97%
4-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 200…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
EINECS: 210-268-2