Ethyl-N-(2-cyanoacetyl)carbamate 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-Cyanoacetylurethane
OR42047-1G OR42047-25G OR42047-5G
APOSOR42047-1GEA 21.9 EUR
APOSOR42047-1G APOSOR42047-25G APOSOR42047-5G
Ethyl-N-(2-cyanoacetyl)carbamate 98%
Ethyl-N-(2-cyanoacetyl)carbamate
Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00001937
CAS Number: 6629-04-5