4-Benzyloxy-3-methoxyphenylacetic acid 98%
Supplier: Apollo Scientific
Danger
OR42061-1G OR42061-5G
APOSOR42061-1GEA 203 EUR
APOSOR42061-1G APOSOR42061-5G
4-Benzyloxy-3-methoxyphenylacetic acid 98%
4-Benzyloxy-3-methoxyphenylacetic acid
Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00185562
CAS Number: 29973-91-9