3-Amino-5-bromo-2-methoxypyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-methoxypyridin-3-amine
OR42139-100G
APOSOR42139-100GEA 183 EUR
APOSOR42139-100G
3-Amino-5-bromo-2-methoxypyridine 98%
3-Amino-5-bromo-2-methoxypyridine
Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
Melting Pt: 57…58 °C
Storage Temperature: Ambient
MDL Number: MFCD07368887
CAS Number: 884495-39-0