3,5-Dibromoanisole
Supplier: Apollo Scientific
Danger
Synonyms: 1,3-Dibromo-5-methoxybenzene
OR42208-25G OR42208-5G OR42208-1G
APOSOR42208-25GEA 132 EUR
APOSOR42208-25G APOSOR42208-5G APOSOR42208-1G
3,5-Dibromoanisole
3,5-Dibromoanisole
Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Boiling Pt: 124 °C/9.75mmHg
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02258848
CAS Number: 74137-36-3
UN: 2811
ADR: 6.1 III