2-Bromo-5-nitrotoluene
Supplier: Apollo Scientific
Synonyms: 1-Bromo-2-methyl-4-nitrobenzene
OR42209-100G OR42209-25G
APOSOR42209-100GEA 63.8 EUR
APOSOR42209-100G APOSOR42209-25G
2-Bromo-5-nitrotoluene
2-Bromo-5-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007281
CAS Number: 7149-70-4