2,2,6,6-Tetramethyl-3,5-heptanedione 98%
Supplier: Apollo Scientific
Warning
Synonyms: Dipivaloylmethane, TMHD, 2,2,6,6-Tetramethylheptane-3,5-dione
OR42216-100G OR42216-25G OR42216-5G
APOSOR42216-100GEA 71.2 EUR
APOSOR42216-100G APOSOR42216-25G APOSOR42216-5G
2,2,6,6-Tetramethyl-3,5-heptanedione 98%
2,2,6,6-Tetramethyl-3,5-heptanedione
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 72…73 °C/6mmHg
Density: 0.883 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3