2,6-Di-tert-butyl-4-methylpyridine 96%
Supplier: Apollo Scientific
Warning
OR42220-25G OR42220-5G OR42220-100G
APOSOR42220-25GEA 114 EUR
APOSOR42220-25G APOSOR42220-5G APOSOR42220-100G
2,6-Di-tert-butyl-4-methylpyridine 96%
2,6-Di-tert-butyl-4-methylpyridine
Formula: C₁₄H₂₃N
Boiling Pt: 233 °C
Melting Pt: 33…36 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00006305
CAS Number: 38222-83-2