2-(3,4-Dimethoxyphenyl)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol
OR42222-100G OR42222-25G OR42222-5G
APOSOR42222-100GEA 154 EUR
APOSOR42222-100G APOSOR42222-25G APOSOR42222-5G
2-(3,4-Dimethoxyphenyl)ethanol
2-(3,4-Dimethoxyphenyl)ethanol
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 172…174 °C/17mmHg
Melting Pt: 44…48 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002894
CAS Number: 7417-21-2
EINECS: 231-032-5