Ethylurea

Supplier: Apollo Scientific
Warning

Synonyms: 1-Ethylurea, N-Ethylurea

OR42229-100G OR42229-25G
APOSOR42229-100GEA 267 EUR
APOSOR42229-100G APOSOR42229-25G
Ethylurea
Ethylurea
Formula: C₃H₈N₂O
Melting Pt: 87…90 °C
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR