1-Bromo-2-ethynylbenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2'-Bromophenylacetylene, 2-Bromophenylacetylene
OR42231-1G OR42231-5G OR42231-25G
APOSOR42231-1GEA 36.4 EUR
APOSOR42231-1G APOSOR42231-5G APOSOR42231-25G
1-Bromo-2-ethynylbenzene 98%
1-Bromo-2-ethynylbenzene
Formula: C₈H₅Br
MW: 181.03 g/mol
Boiling Pt: 92…93 °C/20mmHg
Melting Pt: 52
Density: 1.443 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD01863358
CAS Number: 766-46-1