1-Iodo-2-nitrobenzene
Supplier: Apollo Scientific
Warning
OR4227-100G OR4227-25G
APOSOR4227-100GEA 72.5 EUR
APOSOR4227-100G APOSOR4227-25G
1-Iodo-2-nitrobenzene
1-Iodo-2-nitrobenzene
Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 288…289 °C
Melting Pt: 49…51 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007088
CAS Number: 609-73-4
EINECS: 210-200-1