2-Chloro-3-nitrothiophene 98%
Supplier: Apollo Scientific
Warning
OR4235-1G OR4235-5G OR4235-250MG
APOSOR4235-1GEA 58 EUR
APOSOR4235-1G APOSOR4235-5G APOSOR4235-250MG
2-Chloro-3-nitrothiophene 98%
2-Chloro-3-nitrothiophene
Formula: C₄H₂ClNO₂S
MW: 163.58 g/mol
Melting Pt: 51…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00052159
CAS Number: 5330-98-3